The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006623.3 | NP_006614.2 | 10 - 11 | 1441 | 85 |
XM_011541226.1 | XP_011539528.1 | 12 - 13 | 1722 | 85 | |
XM_011541227.2 | XP_011539529.1 | 12 - 13 | 1427 | 85 | |
XM_011541228.1 | XP_011539530.1 | 12 - 13 | 1340 | 85 | |
XM_011541231.2 | XP_011539533.1 | 12 - 13 | 1544 | 85 | |
GenBank mRNA | AF171235.1 | - | 3 - 4 | 305 | 85 |
AF171236.1 | - | 7 - 8 | 831 | 85 | |
AF171237.1 | - | 10 - 11 | 1287 | 85 | |
AK129762.1 | - | 2 - 3 | 1191 | 85 | |
AK309662.1 | - | 12 - 13 | 1635 | 85 | |
AK315360.1 | - | 10 - 11 | 1362 | 85 | |
BC000303.2 | - | 10 - 11 | 1281 | 85 | |
BC001349.2 | - | 10 - 11 | 1276 | 85 | |
BC011262.1 | - | 10 - 11 | 1350 | 85 | |
BC023235.1 | - | 10 - 11 | 1254 | 85 | |
BC032110.2 | - | 4 - 5 | 582 | 85 | |
CR456795.1 | - | 10 - 11 | 1214 | 85 | |
FJ224347.1 | - | - | 1906 | 85 |