The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006623.3 | NP_006614.2 | 8 - 9 | 1174 | 78 |
XM_011541226.1 | XP_011539528.1 | 10 - 11 | 1455 | 78 | |
XM_011541227.2 | XP_011539529.1 | 10 - 11 | 1160 | 78 | |
XM_011541228.1 | XP_011539530.1 | 10 - 11 | 1073 | 78 | |
XM_011541231.2 | XP_011539533.1 | 10 - 11 | 1277 | 78 | |
GenBank mRNA | AF171236.1 | - | 5 - 6 | 564 | 78 |
AF171237.1 | - | 8 - 9 | 1020 | 78 | |
AK093306.1 | - | 7 - 8 | 2352 | 78 | |
AK309662.1 | - | 10 - 11 | 1368 | 78 | |
AK315360.1 | - | 8 - 9 | 1095 | 78 | |
BC000303.2 | - | 8 - 9 | 1014 | 78 | |
BC001349.2 | - | 8 - 9 | 1009 | 78 | |
BC011262.1 | - | 8 - 9 | 1083 | 78 | |
BC023235.1 | - | 8 - 9 | 987 | 78 | |
BC032110.2 | - | 2 - 3 | 315 | 78 | |
CR456795.1 | - | 8 - 9 | 947 | 78 | |
FJ224347.1 | - | - | 1639 | 78 |