The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032515.4 | NP_115904.1 | 1 - 2 | 259 | 99 |
XM_011511697.2 | XP_011509999.1 | 1 - 2 | 85 | 99 | |
XM_017004775.1 | XP_016860264.1 | 1 - 2 | 167 | 99 | |
GenBank mRNA | AF174487.1 | - | 1 - 2 | 108 | 99 |
BC006203.2 | - | 1 - 2 | 83 | 99 | |
BI550157.1 | - | 1 - 2 | 250 | 99 | |
BX460019.2 | - | 1 - 2 | 259 | 99 |