The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032016.3 | NP_114405.1 | 5 - 6 | 704 | 96 |
XM_005249880.1 | XP_005249937.1 | 6 - 7 | 718 | 96 | |
XM_011515572.1 | XP_011513874.1 | 5 - 6 | 639 | 96 | |
XM_017012692.1 | XP_016868181.1 | 5 - 6 | 737 | 96 | |
XM_017012693.1 | XP_016868182.1 | 6 - 7 | 668 | 96 | |
XM_017012694.1 | XP_016868183.1 | 5 - 6 | 699 | 96 | |
GenBank mRNA | AJ492267.1 | - | 5 - 6 | 579 | 96 |
AK315404.1 | - | 5 - 6 | 673 | 96 | |
AY358645.1 | - | 5 - 6 | 530 | 96 | |
BC003074.2 | - | 4 - 5 | 489 | 96 | |
BC005959.1 | - | 5 - 6 | 524 | 96 |