Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- IL12RB1
- Entrez Gene ID
-
3594
- Gene Name
- interleukin 12 receptor subunit beta 1
- Gene Aliases
-
CD212, IL-12R-BETA1, IL12RB, IMD30
- Chromosome Location
-
Chr.19: 18058994 - 18099027 on Build GRCh38
View Full Product Details