The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001260504.1 | NP_001247433.1 | 12 - 13 | 1984 | 74 |
NM_001260505.1 | NP_001247434.1 | 12 - 13 | 1708 | 74 | |
NM_031414.4 | NP_113602.2 | 12 - 13 | 1708 | 74 | |
NM_032944.3 | NP_116562.2 | 12 - 13 | 1952 | 74 | |
NR_048542.1 | 12 - 13 | 1700 | 74 | ||
XM_006715758.3 | XP_006715821.1 | 10 - 11 | 1088 | 74 | |
XM_011515449.2 | XP_011513751.2 | 12 - 13 | 1634 | 74 | |
XM_011515450.1 | XP_011513752.1 | 12 - 13 | 1955 | 74 | |
XM_011515452.2 | XP_011513754.2 | 10 - 11 | 1101 | 74 | |
GenBank mRNA | AF285599.1 | - | 12 - 13 | 1604 | 74 |
AF332194.1 | - | 12 - 13 | 1987 | 74 | |
AK057271.1 | - | 12 - 13 | 1656 | 74 | |
AK131214.1 | - | 3 - 4 | 410 | 74 | |
AK292474.1 | - | 13 - 14 | 1600 | 74 | |
AK302689.1 | - | 12 - 13 | 1600 | 74 | |
BC033018.1 | - | 12 - 13 | 1602 | 74 | |
BC059374.1 | - | 12 - 13 | 1615 | 74 | |
DA533364.1 | - | 4 - 5 | 486 | 74 |