Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CLN6
- Entrez Gene ID
-
54982
- Gene Name
- ceroid-lipofuscinosis, neuronal 6, late infantile, variant
- Gene Aliases
-
CLN4A, HsT18960, nclf
- Chromosome Location
-
Chr.15: 68206992 - 68229742 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
KU178674.1 |
- |
3 - 4 |
297 |
80 |
View Full Product Details