Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CARD14
Entrez Gene ID
79092
Gene Name
caspase recruitment domain family member 14
Gene Aliases
BIMP2, CARMA2, PRP, PSORS2, PSS1
Chromosome Location
Chr.17: 80169992 - 80209331 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257970.1 NP_001244899.1 12 - 13 1823 68
NM_024110.4 NP_077015.2 12 - 13 1823 68
NM_052819.2 NP_438170.1 9 - 10 1104 68
NR_047566.1 13 - 14 1820 68
XM_011525212.1 XP_011523514.1 14 - 15 2344 68
XM_011525213.1 XP_011523515.1 14 - 15 2086 68
XM_011525215.1 XP_011523517.1 13 - 14 2146 68
XM_011525216.1 XP_011523518.1 13 - 14 1846 68
XM_011525217.1 XP_011523519.1 13 - 14 1902 68
XM_011525218.2 XP_011523520.1 13 - 14 1885 68
XM_017025057.1 XP_016880546.1 14 - 15 2341 68
GenBank mRNA AF322642.1 - 12 - 13 1861 68
AK091123.1 - 13 - 14 1778 68
AK296762.1 - 9 - 10 1044 68
AX746790.1 - 13 - 14 1778 68
AY032927.1 - 11 - 12 1658 68
BC001326.1 - 9 - 10 1104 68
BC018142.1 - 12 - 13 1832 68
EU652409.1 - 11 - 12 1655 68

View Full Product Details