The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257970.1 | NP_001244899.1 | 13 - 14 | 2018 | 70 |
NM_024110.4 | NP_077015.2 | 13 - 14 | 2018 | 70 | |
NR_047566.1 | 14 - 15 | 2015 | 70 | ||
XM_011525212.1 | XP_011523514.1 | 15 - 16 | 2539 | 70 | |
XM_011525213.1 | XP_011523515.1 | 15 - 16 | 2281 | 70 | |
XM_011525215.1 | XP_011523517.1 | 14 - 15 | 2341 | 70 | |
XM_011525216.1 | XP_011523518.1 | 14 - 15 | 2041 | 70 | |
XM_011525217.1 | XP_011523519.1 | 14 - 15 | 2097 | 70 | |
XM_011525218.2 | XP_011523520.1 | 14 - 15 | 2080 | 70 | |
XM_017025057.1 | XP_016880546.1 | 15 - 16 | 2536 | 70 | |
GenBank mRNA | AF322642.1 | - | 13 - 14 | 2056 | 70 |
AK091123.1 | - | 14 - 15 | 1973 | 70 | |
AX746790.1 | - | 14 - 15 | 1973 | 70 | |
AY032927.1 | - | 12 - 13 | 1853 | 70 | |
BC018142.1 | - | 13 - 14 | 2027 | 70 | |
EU652409.1 | - | 12 - 13 | 1850 | 70 |