The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257970.1 | NP_001244899.1 | 14 - 15 | 2145 | 79 |
NM_024110.4 | NP_077015.2 | 14 - 15 | 2145 | 79 | |
NR_047566.1 | 15 - 16 | 2142 | 79 | ||
XM_011525212.1 | XP_011523514.1 | 16 - 17 | 2666 | 79 | |
XM_011525213.1 | XP_011523515.1 | 16 - 17 | 2408 | 79 | |
XM_011525215.1 | XP_011523517.1 | 15 - 16 | 2468 | 79 | |
XM_011525216.1 | XP_011523518.1 | 15 - 16 | 2168 | 79 | |
XM_011525217.1 | XP_011523519.1 | 15 - 16 | 2224 | 79 | |
XM_011525218.2 | XP_011523520.1 | 15 - 16 | 2207 | 79 | |
XM_017025057.1 | XP_016880546.1 | 16 - 17 | 2663 | 79 | |
GenBank mRNA | AF322642.1 | - | 14 - 15 | 2183 | 79 |
AK091123.1 | - | 15 - 16 | 2100 | 79 | |
AX746790.1 | - | 15 - 16 | 2100 | 79 | |
AY032927.1 | - | 13 - 14 | 1980 | 79 | |
BC018142.1 | - | 14 - 15 | 2154 | 79 | |
EU652409.1 | - | 13 - 14 | 1977 | 79 |