The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002615.5 | NP_002606.3 | 3 - 4 | 444 | 89 |
XM_017024754.1 | XP_016880243.1 | 3 - 4 | 364 | 89 | |
GenBank mRNA | AB593011.1 | - | 3 - 4 | 349 | 89 |
AB593012.1 | - | 3 - 4 | 346 | 89 | |
AB593013.1 | - | 3 - 4 | 346 | 89 | |
AF400442.1 | - | 3 - 4 | 360 | 89 | |
AK315344.1 | - | 3 - 4 | 391 | 88 | |
AY513280.1 | - | 3 - 4 | 336 | 89 | |
BC000522.1 | - | 3 - 4 | 339 | 89 | |
BC013984.2 | - | 3 - 4 | 343 | 89 | |
BT007222.1 | - | 2 - 3 | 287 | 89 | |
CA405781.1 | - | 3 - 4 | 394 | 89 | |
M90439.1 | - | 1 - 2 | 112 | 89 |