The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175854.7 | NP_787050.6 | 12 - 13 | 1940 | 68 |
XM_005266332.3 | XP_005266389.1 | 12 - 13 | 1987 | 68 | |
XM_005266333.3 | XP_005266390.1 | 11 - 12 | 1626 | 68 | |
XM_005266334.1 | XP_005266391.1 | 12 - 13 | 1468 | 68 | |
XM_011535032.2 | XP_011533334.1 | 12 - 13 | 1934 | 68 | |
XM_011535033.2 | XP_011533335.1 | 12 - 13 | 1934 | 68 | |
XM_011535034.2 | XP_011533336.1 | 12 - 13 | 1708 | 68 | |
XM_017020528.1 | XP_016876017.1 | 12 - 13 | 1934 | 68 | |
XM_017020529.1 | XP_016876018.1 | 13 - 14 | 1480 | 68 | |
XM_017020530.1 | XP_016876019.1 | 12 - 13 | 1993 | 68 | |
XM_017020531.1 | XP_016876020.1 | 9 - 10 | 1280 | 68 | |
XM_017020532.1 | XP_016876021.1 | 12 - 13 | 1438 | 68 | |
XM_017020533.1 | XP_016876022.1 | 10 - 11 | 1571 | 68 | |
GenBank mRNA | AB107584.1 | - | 10 - 11 | 1188 | 68 |
AB109552.1 | - | 11 - 12 | 1350 | 68 | |
AK131556.1 | - | 6 - 7 | 578 | 68 | |
AK296435.1 | - | 12 - 13 | 1940 | 68 | |
BC024318.1 | - | 5 - 6 | 454 | 68 | |
BC128179.1 | - | 12 - 13 | 1468 | 68 | |
BC128180.1 | - | 10 - 11 | 1188 | 68 |