Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALOXE3
Entrez Gene ID
59344
Gene Name
arachidonate lipoxygenase 3
Gene Aliases
ARCI3, E-LOX, eLOX-3, eLOX3
Chromosome Location
Chr.17: 8095900 - 8118916 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001165960.1 NP_001159432.1 4 - 5 968 75
NM_021628.2 NP_067641.2 4 - 5 804 75
XM_017024921.1 XP_016880410.1 3 - 4 747 75
XM_017024922.1 XP_016880411.1 3 - 4 747 75
XM_017024923.1 XP_016880412.1 3 - 4 747 75
XM_017024924.1 XP_016880413.1 3 - 4 747 75
XM_017024925.1 XP_016880414.1 3 - 4 747 75
GenBank mRNA AF182218.1 - 3 - 4 635 75
AK292732.1 - 3 - 4 707 75
AK296416.1 - 4 - 5 869 75
AK299742.1 - 4 - 5 911 75
AK313677.1 - 4 - 5 768 75
BC101938.1 - 3 - 4 656 75
BC101939.1 - 3 - 4 656 75
BC103508.1 - 3 - 4 656 75
BC104724.1 - 3 - 4 656 75

View Full Product Details