The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165960.1 | NP_001159432.1 | 8 - 9 | 1488 | 67 |
NM_021628.2 | NP_067641.2 | 8 - 9 | 1324 | 67 | |
XM_017024921.1 | XP_016880410.1 | 7 - 8 | 1267 | 67 | |
XM_017024922.1 | XP_016880411.1 | 7 - 8 | 1267 | 67 | |
XM_017024923.1 | XP_016880412.1 | 7 - 8 | 1267 | 67 | |
XM_017024924.1 | XP_016880413.1 | 7 - 8 | 1267 | 67 | |
XM_017024925.1 | XP_016880414.1 | 7 - 8 | 1267 | 67 | |
GenBank mRNA | AF182218.1 | - | 7 - 8 | 1155 | 67 |
AJ269499.1 | - | 7 - 8 | 1453 | 67 | |
AK292732.1 | - | 7 - 8 | 1227 | 67 | |
AK296416.1 | - | 8 - 9 | 1389 | 67 | |
AK299742.1 | - | 8 - 9 | 1431 | 67 | |
AK313677.1 | - | 8 - 9 | 1288 | 67 | |
BC101938.1 | - | 7 - 8 | 1176 | 67 | |
BC101939.1 | - | 7 - 8 | 1176 | 67 | |
BC103508.1 | - | 7 - 8 | 1176 | 67 | |
BC104724.1 | - | 7 - 8 | 1176 | 67 |