The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024577.3 | NP_078853.2 | 10 - 11 | 1331 | 66 |
GenBank mRNA | AB075865.1 | - | 11 - 12 | 1273 | 66 |
AK127241.1 | - | 6 - 7 | 708 | 66 | |
AK127248.1 | - | 11 - 12 | 1338 | 66 | |
AK300420.1 | - | 11 - 12 | 1287 | 66 | |
AY341075.1 | - | 10 - 11 | 1281 | 66 | |
BC113879.1 | - | 10 - 11 | 1246 | 66 | |
BC114486.1 | - | 10 - 11 | 1225 | 66 |