The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005847.4 | NP_005838.3 | 11 - 12 | 1405 | 94 |
NM_152685.3 | NP_689898.2 | 11 - 12 | 1417 | 94 | |
XM_005272148.3 | XP_005272205.3 | 12 - 13 | 1542 | 94 | |
XM_005272149.3 | XP_005272206.3 | 12 - 13 | 1530 | 94 | |
XM_006714741.2 | XP_006714804.2 | 12 - 13 | 1542 | 94 | |
XM_011543765.2 | XP_011542067.1 | 12 - 13 | 1542 | 94 | |
XM_011543766.1 | XP_011542068.1 | 11 - 12 | 1323 | 94 | |
XM_011543767.1 | XP_011542069.1 | 10 - 11 | 1227 | 94 | |
XM_011543768.1 | XP_011542070.1 | 11 - 12 | 1447 | 94 | |
XM_011543769.1 | XP_011542071.1 | 10 - 11 | 1202 | 94 | |
GenBank mRNA | AF098277.1 | - | 11 - 12 | 1355 | 94 |
AF170911.1 | - | 11 - 12 | 1339 | 94 | |
AJ250807.1 | - | 11 - 12 | 1378 | 94 | |
AJ269477.1 | - | 11 - 12 | 1308 | 94 | |
AK092655.1 | - | 10 - 11 | 1229 | 94 | |
AK290789.1 | - | 11 - 12 | 1338 | 94 | |
AK298337.1 | - | 10 - 11 | 1238 | 94 | |
BC050261.1 | - | 11 - 12 | 1359 | 94 |