Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC23A1
Entrez Gene ID
9963
Gene Name
solute carrier family 23 member 1
Gene Aliases
SLC23A2, SVCT1, YSPL3
Chromosome Location
Chr.5: 139367196 - 139384553 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005847.4 NP_005838.3 1 - 2 136 112
NM_152685.3 NP_689898.2 1 - 2 136 112
XM_005272148.3 XP_005272205.3 2 - 3 261 112
XM_005272149.3 XP_005272206.3 2 - 3 261 112
XM_006714741.2 XP_006714804.2 2 - 3 261 112
XM_011543765.2 XP_011542067.1 2 - 3 261 112
XM_011543766.1 XP_011542068.1 2 - 3 261 112
XM_011543767.1 XP_011542069.1 2 - 3 261 112
XM_011543768.1 XP_011542070.1 1 - 2 87 112
XM_011543769.1 XP_011542071.1 2 - 3 261 112
GenBank mRNA AF098277.1 - 1 - 2 86 112
AF170911.1 - 1 - 2 112
AJ250807.1 - 1 - 2 109 112
AK290789.1 - 1 - 2 57 112
AK298337.1 - 1 - 2 96 112
AK300935.1 - 1 - 2 83 112
BC019225.1 - 1 - 2 101 112
BC050261.1 - 1 - 2 78 112
DA385445.1 - 1 - 2 137 112

View Full Product Details