The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005262357.2 | XP_005262414.1 | 1 - 2 | 237 | 64 |
XM_005262361.2 | XP_005262418.1 | 1 - 2 | 184 | 64 | |
XM_017029235.1 | XP_016884724.1 | 1 - 2 | 132 | 64 | |
GenBank mRNA | AK098737.1 | - | 1 - 2 | 191 | 64 |
AK313559.1 | - | 1 - 2 | 41 | 64 | |
DA534452.1 | - | 1 - 2 | 238 | 64 |