The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320866.1 | NP_001307795.1 | 11 - 12 | 1189 | 76 |
NM_001721.6 | NP_001712.1 | 11 - 12 | 1130 | 76 | |
NM_203281.2 | NP_975010.1 | 11 - 12 | 1192 | 76 | |
XM_017029752.1 | XP_016885241.1 | 11 - 12 | 1162 | 76 | |
GenBank mRNA | AF045459.1 | - | 11 - 12 | 1206 | 76 |
AK291824.1 | - | 11 - 12 | 1162 | 76 | |
AK313568.1 | - | 11 - 12 | 1125 | 76 | |
BC016652.2 | - | 11 - 12 | 1147 | 76 | |
X83107.1 | - | 11 - 12 | 1052 | 76 |