Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCDHGA11
Entrez Gene ID
56105
Gene Name
protocadherin gamma subfamily A, 11
Gene Aliases
PCDH-GAMMA-A11
Chromosome Location
Chr.5: 141420970 - 141512979 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018914.2 NP_061737.1 1 - 2 2691 97
GenBank mRNA AF152320.1 - 1 - 2 2433 97
BC136781.1 - 1 - 2 2466 97
BC171774.1 - 1 - 2 2466 97

View Full Product Details