The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127178.2 | NP_001120650.1 | 2 - 3 | 493 | 77 |
NM_001289051.1 | NP_001275980.1 | 2 - 3 | 302 | 77 | |
NM_001289053.1 | NP_001275982.1 | 2 - 3 | 1051 | 77 | |
NM_001289057.1 | NP_001275986.1 | 2 - 3 | 729 | 77 | |
NM_017733.4 | NP_060203.3 | 2 - 3 | 493 | 77 | |
NR_110293.1 | 2 - 3 | 493 | 77 | ||
XM_005272284.2 | XP_005272341.1 | 2 - 3 | 727 | 77 | |
XM_011513490.2 | XP_011511792.1 | 2 - 3 | 456 | 77 | |
XM_011513493.1 | XP_011511795.1 | 2 - 3 | 493 | 77 | |
XM_011513494.2 | XP_011511796.1 | 2 - 3 | 493 | 77 | |
XM_017008321.1 | XP_016863810.1 | 2 - 3 | 727 | 77 | |
XM_017008322.1 | XP_016863811.1 | 2 - 3 | 727 | 77 | |
XM_017008323.1 | XP_016863812.1 | 2 - 3 | 727 | 77 | |
XM_017008324.1 | XP_016863813.1 | 2 - 3 | 727 | 77 | |
XM_017008325.1 | XP_016863814.1 | 2 - 3 | 440 | 77 | |
XM_017008326.1 | XP_016863815.1 | 2 - 3 | 493 | 77 | |
GenBank mRNA | AB162713.1 | - | 2 - 3 | 463 | 77 |
AK074715.1 | - | 2 - 3 | 265 | 77 | |
AK074815.1 | - | 2 - 3 | 472 | 77 | |
AK097244.1 | - | 2 - 3 | 690 | 77 | |
AY358538.1 | - | 2 - 3 | 435 | 77 | |
BC050262.1 | - | 2 - 3 | 963 | 77 | |
BC144576.1 | - | 2 - 3 | 450 | 77 |