The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021154.4 | NP_066977.1 | 3 - 4 | 327 | 108 |
NM_058179.3 | NP_478059.1 | 3 - 4 | 327 | 108 | |
GenBank mRNA | AF113132.1 | - | 3 - 4 | 224 | 108 |
AK025460.1 | - | 3 - 4 | 261 | 108 | |
AK295222.1 | - | 3 - 4 | 327 | 108 | |
AK295310.1 | - | 3 - 4 | 275 | 108 | |
AY131232.1 | - | 3 - 4 | 236 | 108 | |
BC000971.1 | - | 3 - 4 | 249 | 108 | |
BC004863.2 | - | 3 - 4 | 258 | 108 | |
BC016645.2 | - | 3 - 4 | 237 | 108 | |
BC018129.1 | - | 3 - 4 | 258 | 108 | |
BT006840.1 | - | 3 - 4 | 191 | 108 | |
DC300904.1 | - | 3 - 4 | 329 | 108 | |
DC332195.1 | - | 3 - 4 | 300 | 108 |