The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165966.1 | NP_001159438.1 | 5 - 6 | 630 | 55 |
NM_031220.3 | NP_112497.2 | 6 - 7 | 738 | 55 | |
XM_011524015.2 | XP_011522317.1 | 6 - 7 | 678 | 55 | |
XM_011524016.2 | XP_011522318.1 | 6 - 7 | 679 | 55 | |
XM_011524017.2 | XP_011522319.1 | 6 - 7 | 680 | 55 | |
GenBank mRNA | AF334586.1 | - | 6 - 7 | 735 | 55 |
AL389994.1 | - | 5 - 6 | 461 | 55 | |
BC128583.1 | - | 6 - 7 | 675 | 55 | |
BC128584.1 | - | 6 - 7 | 675 | 55 |