Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABCA10
Entrez Gene ID
10349
Gene Name
ATP binding cassette subfamily A member 10
Gene Aliases
EST698739
Chromosome Location
Chr.17: 69148007 - 69244815 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080282.3 NP_525021.3 10 - 11 1881 133
GenBank mRNA AL832004.2 - 9 - 10 1389 133
AY028900.1 - 10 - 11 1881 133
AY247065.1 - 10 - 11 1912 133
BC136460.1 - 7 - 8 1086 133

View Full Product Details