Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCA10
Entrez Gene ID
10349
Gene Name
ATP binding cassette subfamily A member 10
Gene Aliases
EST698739
Chromosome Location
Chr.17: 69148007 - 69244815 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080282.3 NP_525021.3 26 - 27 4030 157
GenBank mRNA AK298508.1 - 3 - 4 374 157
AY028900.1 - 26 - 27 4030 157
AY247065.1 - 26 - 27 4061 157
BC136460.1 - 23 - 24 3235 157
BC144124.1 - 23 - 24 3304 157

View Full Product Details