The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080282.3 | NP_525021.3 | 2 - 3 | 570 | 123 |
GenBank mRNA | AK310858.1 | - | 1 - 2 | 100 | 123 |
AL832004.2 | - | 1 - 2 | 78 | 123 | |
AY028900.1 | - | 2 - 3 | 570 | 123 | |
AY247065.1 | - | 2 - 3 | 601 | 123 |