The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080282.3 | NP_525021.3 | 9 - 10 | 1740 | 92 |
GenBank mRNA | AK310858.1 | - | 7 - 8 | 1166 | 92 |
BC136460.1 | - | 6 - 7 | 945 | 92 | |
BC144124.1 | - | 6 - 7 | 945 | 92 |