Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDPCP
Entrez Gene ID
51057
Gene Name
WD repeat containing planar cell polarity effector
Gene Aliases
BBS15, C2orf86, CHDTHP, FRITZ, FRTZ
Chromosome Location
Chr.2: 63120755 - 63840825 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015910.5 NP_056994.3 4 - 5 718 108
XM_005264348.3 XP_005264405.1 4 - 5 758 108
XM_011532881.2 XP_011531183.1 5 - 6 542 108
XM_011532884.2 XP_011531186.1 4 - 5 758 108
XM_011532886.2 XP_011531188.1 4 - 5 758 108
XM_011532887.2 XP_011531189.1 4 - 5 758 108
XM_011532890.2 XP_011531192.1 4 - 5 758 108
XM_011532891.2 XP_011531193.1 5 - 6 743 108
XM_017004251.1 XP_016859740.1 6 - 7 1396 108
XM_017004252.1 XP_016859741.1 5 - 6 542 108
XM_017004253.1 XP_016859742.1 4 - 5 758 108
XM_017004254.1 XP_016859743.1 4 - 5 758 108
GenBank mRNA AK055030.1 - 5 - 6 743 108
AK310290.1 - 4 - 5 385 108
AL833623.1 - 4 - 5 504 108
BX538331.1 - 4 - 5 464 108

View Full Product Details