The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198461.3 | NP_940863.3 | 3 - 4 | 1556 | 89 |
XM_017003491.1 | XP_016858980.1 | 3 - 4 | 1039 | 89 | |
GenBank mRNA | AK127206.1 | - | 3 - 4 | 258 | 89 |
BC101662.1 | - | 2 - 3 | 215 | 89 | |
BC101664.1 | - | 2 - 3 | 215 | 89 | |
BC143468.1 | - | 2 - 3 | 215 | 89 | |
DQ020495.1 | - | 3 - 4 | 1556 | 89 |