The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198559.1 | NP_001185488.1 | 3 - 4 | 435 | 87 |
NM_016544.2 | NP_057628.1 | 3 - 4 | 435 | 87 | |
XM_011532902.1 | XP_011531204.1 | 3 - 4 | 410 | 87 | |
XM_017004269.1 | XP_016859758.1 | 3 - 4 | 466 | 87 | |
XM_017004270.1 | XP_016859759.1 | 3 - 4 | 326 | 87 | |
GenBank mRNA | AF178983.1 | - | 3 - 4 | 269 | 87 |
AK294726.1 | - | 3 - 4 | 219 | 87 | |
AY094594.1 | - | 3 - 4 | 245 | 87 | |
BC034049.1 | - | 3 - 4 | 296 | 87 |