Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NKD2
Entrez Gene ID
85409
Gene Name
naked cuticle homolog 2
Gene Aliases
Naked2
Chromosome Location
Chr.5: 1008845 - 1038812 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271082.1 NP_001258011.1 4 - 5 304 58
NM_033120.3 NP_149111.1 4 - 5 304 58
XM_005248382.3 XP_005248439.1 4 - 5 421 58
XM_005248383.3 XP_005248440.1 4 - 5 421 58
XM_011514150.2 XP_011512452.1 4 - 5 421 58
GenBank mRNA AB062887.1 - 4 - 5 213 58
AF358137.1 - 4 - 5 225 58
BC012176.1 - 4 - 5 304 58

View Full Product Details