The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000937.4 | NP_000928.1 | 14 - 15 | 68 | |
GenBank mRNA | AK304882.1 | - | 13 - 14 | 68 | |
BC107140.2 | - | 2 - 3 | 343 | 68 | |
BC137231.1 | - | 14 - 15 | 68 | ||
X63564.1 | - | 14 - 15 | 68 |