The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000937.4 | NP_000928.1 | 19 - 20 | 3624 | 112 |
GenBank mRNA | AK304882.1 | - | 18 - 19 | 3270 | 112 |
BC137231.1 | - | 19 - 20 | 3440 | 112 | |
X63564.1 | - | 19 - 20 | 3624 | 112 |