Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POLR2A
Entrez Gene ID
5430
Gene Name
RNA polymerase II subunit A
Gene Aliases
POLR2, POLRA, RPB1, RPBh1, RPO2, RPOL2, RpIILS, hRPB220, hsRPB1
Chromosome Location
Chr.17: 7484379 - 7514618 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000937.4 NP_000928.1 23 - 24 4297 55
GenBank mRNA AK293777.1 - 5 - 6 1011 55
AK304882.1 - 22 - 23 3943 55
BC137231.1 - 23 - 24 4113 55
X63564.1 - 23 - 24 4297 55

View Full Product Details