The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000937.4 | NP_000928.1 | 27 - 28 | 4993 | 85 |
GenBank mRNA | AK293777.1 | - | 9 - 10 | 1707 | 85 |
AK304882.1 | - | 26 - 27 | 4639 | 85 | |
BC137231.1 | - | 27 - 28 | 4809 | 85 | |
DA923649.1 | - | 1 - 2 | 101 | 85 |