Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARFIP2
Entrez Gene ID
23647
Gene Name
ADP ribosylation factor interacting protein 2
Gene Aliases
POR1
Chromosome Location
Chr.11: 6474683 - 6481479 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242854.1 NP_001229783.1 1 - 2 250 128
XM_005252840.4 XP_005252897.1 1 - 2 114 128
GenBank mRNA AK300855.1 - 1 - 2 111 128
AK301972.1 - 1 - 2 217 128
AK309198.1 - 1 - 2 114 128
AK311655.1 - 1 - 2 121 128

View Full Product Details