The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257180.1 | NP_001244109.1 | 3 - 4 | 1128 | 67 |
NM_001257181.1 | NP_001244110.1 | 3 - 4 | 759 | 67 | |
NM_006749.4 | NP_006740.1 | 3 - 4 | 1091 | 67 | |
XM_005273613.3 | XP_005273670.1 | 3 - 4 | 866 | 67 | |
XM_005273615.3 | XP_005273672.1 | 3 - 4 | 866 | 67 | |
XM_006716391.3 | XP_006716454.1 | 4 - 5 | 4346 | 67 | |
XM_017013748.1 | XP_016869237.1 | 3 - 4 | 982 | 67 | |
XM_017013750.1 | XP_016869239.1 | 4 - 5 | 1001 | 67 | |
XM_017013751.1 | XP_016869240.1 | 4 - 5 | 875 | 67 | |
XM_017013752.1 | XP_016869241.1 | 4 - 5 | 1117 | 67 | |
GenBank mRNA | AK291202.1 | - | 3 - 4 | 1128 | 67 |
AK314774.1 | - | 3 - 4 | 760 | 67 | |
BC028600.2 | - | 3 - 4 | 803 | 67 | |
L20852.1 | - | 2 - 3 | 677 | 67 |