Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC20A2
Entrez Gene ID
6575
Gene Name
solute carrier family 20 member 2
Gene Aliases
GLVR-2, GLVR2, IBGC1, IBGC3, MLVAR, PIT-2, PIT2, RAM1, Ram-1
Chromosome Location
Chr.8: 42416462 - 42542213 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257180.1 NP_001244109.1 9 - 10 2408 79
NM_001257181.1 NP_001244110.1 9 - 10 2039 79
NM_006749.4 NP_006740.1 9 - 10 2371 79
XM_005273613.3 XP_005273670.1 9 - 10 2146 79
XM_005273615.3 XP_005273672.1 9 - 10 2146 79
XM_006716390.3 XP_006716453.1 8 - 9 2005 79
XM_006716391.3 XP_006716454.1 10 - 11 5626 79
XM_017013748.1 XP_016869237.1 9 - 10 2262 79
XM_017013749.1 XP_016869238.1 8 - 9 5350 79
XM_017013750.1 XP_016869239.1 10 - 11 2281 79
XM_017013751.1 XP_016869240.1 10 - 11 2155 79
XM_017013752.1 XP_016869241.1 10 - 11 2397 79
GenBank mRNA AK291202.1 - 9 - 10 2408 79
AK295952.1 - 5 - 6 1249 79
AK314774.1 - 9 - 10 2040 79
BC028600.2 - 9 - 10 2083 79
L20852.1 - 8 - 9 1957 79

View Full Product Details