The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013302.3 | NP_037434.1 | 15 - 16 | 2252 | 96 |
XM_017023193.1 | XP_016878682.1 | 15 - 16 | 1963 | 96 | |
XM_017023194.1 | XP_016878683.1 | 14 - 15 | 2196 | 96 | |
GenBank mRNA | BC032665.1 | - | 16 - 17 | 1963 | 96 |
U93850.1 | - | 14 - 15 | 1767 | 96 |