The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002186.2 | NP_002177.2 | 7 - 8 | 1065 | 67 |
XM_011531151.2 | XP_011529453.1 | 8 - 9 | 1115 | 67 | |
XM_011531152.2 | XP_011529454.1 | 8 - 9 | 1091 | 67 | |
XM_011531154.2 | XP_011529456.1 | 8 - 9 | 1137 | 67 | |
XM_011531155.2 | XP_011529457.1 | 8 - 9 | 1124 | 67 | |
XM_011531156.2 | XP_011529458.1 | 8 - 9 | 1122 | 67 | |
XM_017029495.1 | XP_016884984.1 | 8 - 9 | 1114 | 67 | |
XM_017029497.1 | XP_016884986.1 | 8 - 9 | 1161 | 67 | |
XM_017029498.1 | XP_016884987.1 | 9 - 10 | 1266 | 67 | |
XM_017029499.1 | XP_016884988.1 | 9 - 10 | 1188 | 67 | |
XM_017029500.1 | XP_016884989.1 | 8 - 9 | 1128 | 67 | |
XM_017029502.1 | XP_016884991.1 | 4 - 5 | 568 | 67 | |
XM_017029503.1 | XP_016884992.1 | 9 - 10 | 1167 | 67 | |
GenBank mRNA | AK303178.1 | - | 8 - 9 | 1129 | 67 |
M84747.1 | - | 7 - 8 | 1065 | 67 |