The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098398.1 | NP_001091868.1 | 30 - 31 | 3683 | 57 |
NM_004371.3 | NP_004362.2 | 30 - 31 | 3656 | 57 | |
GenBank mRNA | AK129830.1 | - | 4 - 5 | 501 | 57 |
BC038447.1 | - | 30 - 31 | 3378 | 57 | |
U24105.1 | - | 31 - 32 | 3728 | 57 |