The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098398.1 | NP_001091868.1 | 2 - 3 | 550 | 71 |
NM_004371.3 | NP_004362.2 | 2 - 3 | 550 | 71 | |
GenBank mRNA | BC038447.1 | - | 2 - 3 | 245 | 71 |
U24105.1 | - | 3 - 4 | 622 | 71 |