The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031717.3 | NP_001026887.1 | 2 - 3 | 273 | 76 |
NM_001077415.2 | NP_001070883.1 | 2 - 3 | 273 | 76 | |
NM_015513.4 | NP_056328.2 | 1 - 2 | 773 | 76 | |
XM_011534108.1 | XP_011532410.1 | 2 - 3 | 258 | 76 | |
XM_017007175.1 | XP_016862664.1 | 2 - 3 | 258 | 76 | |
GenBank mRNA | AF452623.1 | - | 1 - 2 | 534 | 76 |
AK314113.1 | - | 1 - 2 | 717 | 76 | |
AL050275.1 | - | 1 - 2 | 773 | 76 | |
AY358363.1 | - | 2 - 3 | 276 | 76 | |
BC008720.2 | - | 2 - 3 | 258 | 76 | |
BX358821.2 | - | 2 - 3 | 222 | 76 | |
CR457380.1 | - | 1 - 2 | 174 | 76 | |
DN996843.1 | - | 2 - 3 | 275 | 76 |