Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GON4L
Entrez Gene ID
54856
Gene Name
gon-4 like
Gene Aliases
GON-4, GON4, YARP
Chromosome Location
Chr.1: 155749658 - 155859400 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282856.1 NP_001269785.1 25 - 26 5378 56
NM_001282858.1 NP_001269787.1 25 - 26 5430 56
NM_001282860.1 NP_001269789.1 25 - 26 5430 56
XM_005245284.3 XP_005245341.1 25 - 26 5370 56
XM_005245286.3 XP_005245343.1 10 - 11 3181 56
XM_006711393.3 XP_006711456.1 25 - 26 5389 56
XM_006711394.3 XP_006711457.1 25 - 26 5375 56
XM_011509658.2 XP_011507960.1 24 - 25 5270 56
XM_011509659.2 XP_011507961.1 25 - 26 5394 56
GenBank mRNA AB046826.1 - 5 - 6 1821 56
AB195687.1 - 25 - 26 5430 56
AB195688.1 - 26 - 27 4112 56
AK000210.1 - 4 - 5 631 56
AY335490.2 - 25 - 26 5333 56
AY335491.2 - 25 - 26 5378 56
BC064933.1 - 4 - 5 693 56
BC117557.1 - 10 - 11 3193 56

View Full Product Details