Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GON4L
Entrez Gene ID
54856
Gene Name
gon-4 like
Gene Aliases
GON-4, GON4, YARP
Chromosome Location
Chr.1: 155749658 - 155859400 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282856.1 NP_001269785.1 26 - 27 5526 81
NM_001282858.1 NP_001269787.1 26 - 27 5578 81
NM_001282860.1 NP_001269789.1 26 - 27 5578 81
XM_005245284.3 XP_005245341.1 26 - 27 5518 81
XM_005245286.3 XP_005245343.1 11 - 12 3329 81
XM_006711393.3 XP_006711456.1 26 - 27 5537 81
XM_006711394.3 XP_006711457.1 26 - 27 5523 81
XM_011509658.2 XP_011507960.1 25 - 26 5418 81
XM_011509659.2 XP_011507961.1 26 - 27 5542 81
GenBank mRNA AB046826.1 - 6 - 7 1969 81
AB195687.1 - 26 - 27 5578 81
AB195688.1 - 27 - 28 4260 81
AK000210.1 - 5 - 6 779 81
AY335490.2 - 26 - 27 5481 81
AY335491.2 - 26 - 27 5526 81
BC064933.1 - 5 - 6 841 81
BC117557.1 - 11 - 12 3341 81

View Full Product Details