The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001178061.1 | NP_001171532.1 | 13 - 14 | 1597 | 65 |
NM_001178062.1 | NP_001171533.1 | 12 - 13 | 1477 | 65 | |
NM_030913.4 | NP_112175.2 | 13 - 14 | 1597 | 65 | |
XM_005244835.3 | XP_005244892.1 | 13 - 14 | 1601 | 65 | |
XM_017000075.1 | XP_016855564.1 | 13 - 14 | 2008 | 65 | |
XM_017000076.1 | XP_016855565.1 | 13 - 14 | 2018 | 65 | |
XM_017000077.1 | XP_016855566.1 | 14 - 15 | 2930 | 65 | |
XM_017000078.1 | XP_016855567.1 | 12 - 13 | 2597 | 65 | |
XM_017000079.1 | XP_016855568.1 | 14 - 15 | 2930 | 65 | |
XM_017000080.1 | XP_016855569.1 | 11 - 12 | 2477 | 65 | |
XM_017000081.1 | XP_016855570.1 | 13 - 14 | 2810 | 65 | |
XM_017000082.1 | XP_016855571.1 | 6 - 7 | 709 | 65 | |
GenBank mRNA | AB022434.1 | - | 13 - 14 | 1448 | 65 |
AB058772.1 | - | 13 - 14 | 2953 | 65 | |
AF339152.1 | - | 13 - 14 | 1390 | 65 | |
AF339153.1 | - | 12 - 13 | 1270 | 65 | |
AF339154.1 | - | 13 - 14 | 1390 | 65 | |
AK302862.1 | - | 13 - 14 | 1564 | 65 |