The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001178061.1 | NP_001171532.1 | 17 - 18 | 2048 | 70 |
NM_001178062.1 | NP_001171533.1 | 16 - 17 | 1928 | 70 | |
NM_030913.4 | NP_112175.2 | 17 - 18 | 2048 | 70 | |
XM_005244835.3 | XP_005244892.1 | 17 - 18 | 2052 | 70 | |
XM_017000075.1 | XP_016855564.1 | 17 - 18 | 2459 | 70 | |
XM_017000076.1 | XP_016855565.1 | 17 - 18 | 2469 | 70 | |
XM_017000077.1 | XP_016855566.1 | 18 - 19 | 3381 | 70 | |
XM_017000078.1 | XP_016855567.1 | 16 - 17 | 3048 | 70 | |
XM_017000079.1 | XP_016855568.1 | 18 - 19 | 3381 | 70 | |
XM_017000080.1 | XP_016855569.1 | 15 - 16 | 2928 | 70 | |
XM_017000081.1 | XP_016855570.1 | 17 - 18 | 3261 | 70 | |
XM_017000082.1 | XP_016855571.1 | 10 - 11 | 1160 | 70 | |
GenBank mRNA | AB022434.1 | - | 17 - 18 | 1899 | 70 |
AB058772.1 | - | 17 - 18 | 3404 | 70 | |
AF055020.1 | - | 3 - 4 | 262 | 70 | |
AF339152.1 | - | 17 - 18 | 1841 | 70 | |
AF339153.1 | - | 16 - 17 | 1721 | 70 | |
AF339154.1 | - | 17 - 18 | 1841 | 70 | |
AK302862.1 | - | 17 - 18 | 2015 | 70 | |
BE253393.1 | - | 3 - 4 | 206 | 70 |