The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001178061.1 | NP_001171532.1 | 4 - 5 | 571 | 61 |
NM_001178062.1 | NP_001171533.1 | 4 - 5 | 571 | 61 | |
NM_030913.4 | NP_112175.2 | 4 - 5 | 571 | 61 | |
XM_005244835.3 | XP_005244892.1 | 4 - 5 | 575 | 61 | |
XM_017000075.1 | XP_016855564.1 | 4 - 5 | 982 | 61 | |
XM_017000076.1 | XP_016855565.1 | 4 - 5 | 992 | 61 | |
XM_017000077.1 | XP_016855566.1 | 5 - 6 | 1904 | 61 | |
XM_017000078.1 | XP_016855567.1 | 3 - 4 | 1571 | 61 | |
XM_017000079.1 | XP_016855568.1 | 5 - 6 | 1904 | 61 | |
XM_017000080.1 | XP_016855569.1 | 3 - 4 | 1571 | 61 | |
XM_017000081.1 | XP_016855570.1 | 5 - 6 | 1904 | 61 | |
GenBank mRNA | AB022434.1 | - | 4 - 5 | 422 | 61 |
AB058772.1 | - | 4 - 5 | 1927 | 61 | |
AF339152.1 | - | 4 - 5 | 364 | 61 | |
AF339153.1 | - | 4 - 5 | 364 | 61 | |
AF339154.1 | - | 4 - 5 | 364 | 61 | |
AK302862.1 | - | 4 - 5 | 538 | 61 | |
AK311613.1 | - | 3 - 4 | 366 | 61 | |
BC114521.1 | - | 3 - 4 | 311 | 61 | |
BC114623.1 | - | 3 - 4 | 311 | 61 |