The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016155.5 | NP_057239.4 | 3 - 4 | 525 | 68 |
XM_011538355.2 | XP_011536657.1 | 3 - 4 | 379 | 68 | |
XM_011538356.2 | XP_011536658.1 | 3 - 4 | 375 | 68 | |
XM_011538357.2 | XP_011536659.1 | 3 - 4 | 324 | 68 | |
XM_017019307.1 | XP_016874796.1 | 3 - 4 | 504 | 68 | |
GenBank mRNA | AB021225.2 | - | 3 - 4 | 531 | 68 |
BC040507.1 | - | 3 - 4 | 517 | 68 | |
BC045610.1 | - | 3 - 4 | 534 | 68 | |
CA489162.1 | - | 1 - 2 | 136 | 68 | |
X89576.1 | - | 2 - 3 | 403 | 68 |