The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015906.3 | NP_056990.3 | 16 - 17 | 2978 | 156 |
NM_033020.2 | NP_148980.2 | 16 - 17 | 2978 | 156 | |
XM_005270936.3 | XP_005270993.1 | 17 - 18 | 2971 | 156 | |
XM_005270937.3 | XP_005270994.1 | 17 - 18 | 2971 | 156 | |
XM_011541568.2 | XP_011539870.1 | 17 - 18 | 2968 | 156 | |
XM_017001452.1 | XP_016856941.1 | 14 - 15 | 2253 | 156 | |
XM_017001453.1 | XP_016856942.1 | 14 - 15 | 2253 | 156 | |
GenBank mRNA | AB029036.1 | - | 16 - 17 | 2907 | 156 |
AF119043.1 | - | 16 - 17 | 2978 | 156 | |
AF220136.1 | - | 16 - 17 | 2978 | 156 | |
AF220137.1 | - | 16 - 17 | 2978 | 156 | |
AJ132948.1 | - | 16 - 17 | 2669 | 156 | |
AK023434.1 | - | 14 - 15 | 2191 | 156 | |
AK057487.1 | - | 11 - 12 | 1195 | 156 | |
AK301997.1 | - | 11 - 12 | 1656 | 156 |